FCS3® Non-Heating Chamber Set, Bioptechs®
# de catalogue 10200-112
Fournisseur: BIOPTECHS INC.
Spécifications
- Description:FCS3® Non-Heating Chamber Set
- Cat. no.:10200-112
- Supplier No.:210603193NH
Spécifications
A propos de cet article
Compatible, microscopic flow cell with a user definable specimen area for upright microscopes.
- Unparalleled, uniform temperature control across entire specimen area
- Near laminar flow perfusion
- User definable flow characteristics
- Compatible with all modes of microscopy including high NA/high magnification uses
- Over 20 years of being the foremost chamber in micro-environmental control
- Ideal for both flow and static applications
- A parallel plate, highly N.A.
Certifications: CE certified.
Ordering information: Order includes Chamber only.
Customers Who Bought This Also Bought