FCS2® Temperature Controller, Bioptechs Inc.®
# de catalogue 10200-008
Fournisseur: BIOPTECHS INC.
Spécifications
- Description:FCS2® Temperature Controller
- Cat. no.:10200-008
- Supplier No.:06031920303
Spécifications
A propos de cet article
The FCS2® Temperature Controller is an electronic controller used with the FCS2® chamber to maintain uniform temperature, from ambient to 50 °C.
- Unparalleled, uniform temperature control across entire specimen area
- Compatible with all modes of microscopy including high NA/high magnification uses
Includes intelligent feedback system.
Certifications: CE certified.
Delivery: Controller is supplied with 12 VDC power supply.
Customers Who Bought This Also Bought