FCS2® Chamber, Bioptechs
# de catalogue 10200-006
Fournisseur: BIOPTECHS INC.
Spécifications
- Description:FCS2® Chamber
- Cat. no.:10200-006
- Supplier No.:060319203
Spécifications
A propos de cet article
A parallel plate, high N.A
- Unparalleled, uniform temperature control across entire specimen area
- Near laminar flow perfusion
- User definable flow characteristics
- Compatible with all modes of microscopy including high NA/high magnification uses
- Over 20 years of being the foremost chamber in micro-environmental control
- Ideal for both flow and static applications
compatible, uniformly temperature controlled microscopic flow cell with a user definable specimen area.
Certifications: CE certified.
Ordering information: Chamber only, requires FCS2® Controller.
Customers Who Bought This Also Bought