FCS2® Micro-Perfusion Pumps, Bioptechs Inc.®
Fournisseur: BIOPTECHS INC.
Certificats
A propos de cet article
These single-channel, smooth, analog motor-driven peristaltic pumps are designed specifically for microscopy applications requiring a variety of flow rates.
- Compact
- Usable in single- or dual-channel format
- Precise speed setting feature
- Flow rates of 0.8µL/min. to 18mL/min. (pump dependant)
- Compatible with 1/16"" tubing
- Universal 15VDC adapter accepts 100–240VAC input (power supply has plugs for US, Europe, UK, and Australia)
- 9V battery backup
- Internal battery can function as primary power supply
- Computer interface available
Certifications: CE certified.
Ordering information: Pump requires tubing.
Customers Who Bought This Also Bought