Perfusion/Temperature USB Controller, Bioptechs Inc.®
# de catalogue 10200-002
Fournisseur: BIOPTECHS INC.
Spécifications
- Description:Perfusion/Temperature USB Controller
- Cat. no.:10200-002
- Supplier No.:1316160313
Spécifications
A propos de cet article
A USB interfaced computer program and associated cabling designed to interface micro-perfusion pumps and Bioptechs temperature controllers
- Dual or single micro-perfusion pumps
- Flow profile to reduce dead volume delays
- Temperature and perfusion recording
- Data logging of perfusion and temperature
- Multi flow-rate calibration
- Temperature profiling and cycling
- Saving and reloading settings
- Graphic display of events
The program is compatible with Windows XP, Windows 7, and Windows 8 systems. The program provides an intuitive means of control for perfusion or temperature in time lapse micro-imaging.
Certifications: CE certified.
Ordering information: Software only.
Customers Who Bought This Also Bought