CFCS2® Cooled Closed Chamber System, Bioptechs Inc.®
# de catalogue 10200-026
Fournisseur: BIOPTECHS INC.
Spécifications
- Description:CFCS2® Cooled Closed Chamber System
- Cat. no.:10200-026
- Supplier No.:030603192
Spécifications
A propos de cet article
This modified FCS2® Closed Chamber System includes fluid ports in the base to accommodate the circulation of an external refrigerant.
- Suitable for no flow through high rate flow procedures where a rapid exchange of media is required with low cell surface shear
- Closed system so that bicarbonate CO2 or organic buffers can be employed
- Compatible with 1/16" tubing for perfusion (C-Flex, Tygon, etc.)
- Easily assembled with no tools required
- Unlimited options for internal geometry
- Creates user definable flow characteristics
- Enables near laminar flow
Certifications: CE certified.
Ordering information: System includes Chamber, five Non-Heating Microaqueduct Slides, fifty Coverslips, and 30-piece Gasket Set. This chamber has heating and cooling capabilities, but does not include temperature controller.
Customers Who Bought This Also Bought