tec clean A4 Universal Ultrasonic Cleaning and Degreasing Solution, Elma
Fournisseur: Avantor
Certificats
A propos de cet article
Safe and cost effective Elma cleaning solutions prolong the lifetime of your ultrasonic bath.
- Alkaline ultrasonic cleaning and degreasing concentrate
- Cleans metals, glass, rubber, ceramics and alkaline-resistant plastics
- Removes cutting and machining coolant, grease, oils, soot, carbon, wax, polishing media, fingerprints, and dust
Oil, grease, and wax removed from parts will separate and float on the liquid surface where they can be skimmed off.
Biodegradable Elma tec clean A4 is highly concentrated and should be diluted to 2-5% with water before use. A 10 liter bottle dilutes to 132 gallons of 2% solution.
Spécifications
- Catalog No:
- 10724-600
- Environmentally Preferable:
- Sustainable Packaging, Safer Human and Environmental Health