Oasis® Heating and Cooling System, Bioptechs Inc.®
# de catalogue 10199-782
Fournisseur: BIOPTECHS INC.
Spécifications
- Description:Oasis® Heating and Cooling System
- Cat. no.:10199-782
- Supplier No.:15160
Spécifications
A propos de cet article
The Oasis® Heating and Cooling System is a Peltier-driven heating and cooling system that circulates temperature-regulated fluid through accessory devices used on a microscope.
- Lightweight and portable
- User-friendly interface
- Universal power input
- Low power consumption
- Solid-State technology (no compressor)
- Environmentally friendly
- Leak-free disconnects
- Quiet operation
- Serial communication
- Non-volatile memory set temperature
- Wide temperature range of operation
Certifications: CE certified.
Ordering information: The Oasis® Heating and Cooling System requires tubing and cooling adapter for specific cooling.
Customers Who Bought This Also Bought