Blank Gaskets, Bioptechs Inc.®
Supplier: BIOPTECHS INC.
Certificates
About this item
These flat, 40 mm diameter silicone sheets are available in varied thicknesses, and can be modified by the user to suit specific protocols.
- Unlimited options for internal geometry
- Creates user definable flow characteristics
- Enables near laminar flow
Ordering information: Each order includes 5 gaskets.
Customers Who Bought This Also Bought