Jomac® Cut and Sewn Heavy Weight Terry Cloth Gloves, Loop Out, Wells Lamont
Supplier: Wells Lamont
Certificates
About this item
Jomac cut and sewn heavy weight gloves protect against up to 325 °F.
- Natural white
- Loop-out terry cloth
- Knit-wrist cuff
- Ambidextrous
- Heavy weight
Users can clean the ambidextrous gloves in the washing machine.
Ordering information: Gloves are sold by the dozen pairs.
Caution: The degree of heat resistance a glove may offer is directly related to the duration of exposure and weight of the object being handled.
Specifications
- Catalog No:
- 80091-248
- 80091-236
- Ambidextrous/Hand-specific:
- Ambidextrous
- Ambidextrous
- Cuff style:
- Knit Wrist
- Knit Wrist