ADA Remote Control, Labconco
# de catalogue 10769-376
Fournisseur: Labconco
Spécifications
- Description:ADA Remote Control
- Cat. no.:10769-376
- Supplier No.:9579700
Spécifications
A propos de cet article
Allows persons in wheelchairs and/or shorter individuals access to the fan and light switches through a wireless remote control.
WxDxH: 2 x 4 x 1"
Weight: 1 lb.
Ordering information: For use with Protector® Filtered Fume Hoods. Includes 1 lithium battery.
Customers Who Bought This Also Bought