gGuard® communication software, Labconco
# de catalogue 10769-424
Fournisseur: Labconco
Product available on GSA Advantage®
Spécifications
- Description:gGuard® Communication Software, 2 x 5 x 1/2"
- Cat. no.:10769-424
- Supplier No.:9581100
Spécifications
A propos de cet article
Automatically collects and records hood data, alerts and alarms on a customer-supplied personal computer
Monitors, manages and provides data via email to a designated facility manager. Provides access to the following: usage authorization, filter usage, Neutrodine Filter saturation detection, filter identification, sash position, blower speed, laboratory air pollution detection, temperature and usage statistics. Software available on CD-ROM.
Ordering information: For use with Protector® Filtered Fume Hoods.
PC requirements include Windows 7 or newer operating system; x86 (32 bits)/x64 (64 bits)/ia64; Processor x86 - 1 Gb or more, 512 Mb min x 64 - 1, 4Gb or more; 850 Mb + 5 Gb free memory hard disk for the SQL database (x86 processor) or 2 Gb + 5 Gb free memory hard disk for the SQL database (x64 processor); 32 bits graphic card, 10/100/1000 Mbps ethernet card; and CD-ROM player.
PC requirements include Windows 7 or newer operating system; x86 (32 bits)/x64 (64 bits)/ia64; Processor x86 - 1 Gb or more, 512 Mb min x 64 - 1, 4Gb or more; 850 Mb + 5 Gb free memory hard disk for the SQL database (x86 processor) or 2 Gb + 5 Gb free memory hard disk for the SQL database (x64 processor); 32 bits graphic card, 10/100/1000 Mbps ethernet card; and CD-ROM player.
Customers Who Bought This Also Bought