Circulation Tubing Kit, Labconco®
# de catalogue 10002-756
Fournisseur: Labconco
Product available on GSA Advantage®
Spécifications
- Length:104 cm (41")
- Tubing I.D.:1.9 cm (³⁄₄")
- Description:Circulation tubing kit, 1.9 cm (3/4") O.D. Fittings
- Cat. no.:10002-756
- Supplier No.:5441808
Spécifications
A propos de cet article
Two each sections of 1.9 cm (³/₄") int.Ø flexible neoprene tubing with 1.9 cm (³/₄") ext.Ø brass compression nut tube fittings on each end.
Tubing kit is desgined for connecting Glove box monitor to Labconco® Glove box and AtmosPure™ Re-Gen gas purifier.
Customers Who Bought This Also Bought