ContactUsLinkComponent
AtmoSense™ Moisture Monitor, Labconco®
AtmoSense™ Moisture Monitor, Labconco®
# de catalogue 10002-754
Fournisseur:  Labconco
Product available on GSA Advantage®
Atmosense oxygen monitor
AtmoSense™ Moisture Monitor, Labconco®
# de catalogue 10002-754
Fournisseur:  Labconco
Product available on GSA Advantage®
Spécifications
  • Description:
    AtmoSense™ Moisture Monitor, 0–12,310 ppm
  • Height:
    111/4"
  • Cat. no.:
    10002-754
  • Electrical:
    115 V
  • Depth:
    17"
  • Width:
    10"
  • Supplier No.:
    5255600
Spécifications
A propos de cet article
The AtmoSense™ Moisture Monitor is designed for general purpose trace moisture detection as low as 0–20ppm
Designed for placement atop transfer chamber of a Labconco® Protector® Controlled Atmosphere, Combination Glove Box, or the Precise® Controlled Atmosphere Glove Box rather than inside. Operates in-line with the AtmosPure™ Re-Gen Gas Purifier. Constructed with a durable glacier white powder-coated housing. Gas inlet and outlet connections are 1.9cm (3/4") brass compression fittings.
An analog output signal of 0–5 volts is provided from –50 to 10°C dew point (64–12,310 ppm). Auto-calibration occurs every 30 minutes and is integrated between 20–1,250ppm.
The device provides a digital LCD readout from 0–12,310ppm moisture (0–52% RH), with resolution accuracy of 5ppm (2°C dew point) measured at 20ppm (–60°C dew point). Includes certificate of initial calibration.
Ordering information: The AtmoSense™ Moisture Monitor must be used with Circulation Tubing Kit 5441808 (sold separately). The AtmoSense™ Moisture Monitor includes 100–115 volt cord with IEC 320 C14 plug for North America/Japan use. With 12 VDC input voltage for operation on 50/60Hz powered from 100, 115, or 208/240 volt AC. Power cords with other plug types are available (sold separately).
Customers Who Bought This Also Bought