Anti-YPYDVPDYA Mouse Polyclonal Antibody (DyLight® 550) [clone: 16B12]
# de catalogue 10065-734
Fournisseur: Columbia Biosciences
Restricted Products: To process your orders without delay, please provide the required business documentation to purchase this product.
To order chemicals, medical devices, or other restricted products please provide ID that includes your business name & shipping address via email [email protected] or fax 484.881.5997 referencing your VWR account number. Acceptable forms of ID are:
- • State issued document with your organization's Federal Tax ID Number
- • State issued document with your organization's Resale Tax ID Number
- • City or County issued Business License
- • State Department of Health Services License
- • Any other ID issued by the State that includes the business name & address
* ATTN: California Customers may require additional documentation as part of the CA Health & Safety Code. Products that fall under this regulation will be placed on a mandatory 21-day hold after documentation is received. VWR will not lift restrictions for residential shipping addresses.
Spécifications
- Antibody type:Tag
- Clonality:Polyclonal
- Clone:16B12
- Host:Mouse
- Isotype:IgG
- Reactivity:Human
- Antigen symbol:YPYDVPDYA
- Conjugation:DyLight® 550
- ELISA:Yes
- Flow cytometry:Yes
- ImmunoChemistry:Yes
- ImmunoFluorescence:Yes
- Size:100 µg
- Form:liquid
- Storage buffer:pH 7.4
- Storage temperature:Store at 4 °C
- Concentration:100 µg at 1mg/ml
- Shipping temperature:4 °C
- Immunogen:synthetic peptide corresponds to HA tag
- Tested applications:High Throughput Screening, Fluorescence Resonance Energy Transfer Assays Targeting Kinase Activity and Flow Cytometry
- Purification:protein G purified
- Cat. no.:10065-734
- Packaging:plastic vials
- Supplier No.:D9-1722
Spécifications
A propos de cet article
Mouse monoclonal [16B12] to HA tag (Dylight 550)
Type:
Antigen: YPYDVPDYA
Clonality: Polyclonal
Clone: 16B12
Conjugation: DyLight® 550
Epitope:
Host: Mouse
Isotype: IgG
Reactivity: Human
Customers Who Bought This Also Bought