VWR® Drawer Mounting Brackets for VWR® Benches
Fournisseur: VWR International
Certificats
A propos de cet article
Drawer/cabinet units, including Contour, can be easily mounted to VWR® benches using these drawer brackets.
- Sleek design
- Hardware included
Ordering information: In order to determine the necessary bracket, select your drawer unit type and bench style/size from the table below. Only the following Contour cabinets can be suspended from VWR® C-leg and 4-leg benches. These include modules CFC03M1918PB, CFC03M1918LPB, CFD00M1918PB, CFD85M1918PB, CFC03M2518PB, CFC03M2518LPB, CFE40M2518PB, CFE40M2518LPB, CFD85M2518PB, and CFD00M2518PB.
Customers Who Bought This Also Bought