Dosing Pumps, Dosi-Pump, Bürkle
Fournisseur: BURKLE INC
Certificats
A propos de cet article
Dosi pump are hand controlled pump used for the controlled product extraction from canisters, bottles, barrels and carboys
- Medium contacting parts made of PP, PE and stainless steel
- Suitable for products of different viscosity
Dosi pump is inserted into container closures with suitable holes. Immersion value represents the value of Immersion depth.
Customers Who Bought This Also Bought