Disposable Samplers, ZoneDispo, Burkle
Catalog # 10769-584
Supplier: BURKLE INC
Specifications
- Ø:25 mm (0.98")
- Volume:100 ml (3.38 oz.)
- Description:HDPE, ZoneDispo, with Three Holes
- Length:500 mm (19.69")
- Cat. no.:10769-584
- Sterile:Sterile
- Supplier No.:5393-4421
Specifications
About this item
ZoneDispo disposable all-layer sampler is used in sampling to collect a representative all-layer sample from three different layers.
- HDPE
- Clean room manufactured
- Optionally sterilised by gamma rays
Suitable for free-flowing powders and small-grain granulates. The collected sample can be secured and transported in the closed sampler.
Customers Who Bought This Also Bought