LaboPlast® and SteriPlast® Bio Samplling Spoons, Bürkle
Fournisseur: BURKLE INC
Certificats
A propos de cet article
These disposable spoons are manufactured from bio-plastic "Green PE" which is entirely made of renewable resources
- "Green PE" is 100% recyclable, made without fossil raw materials
- Available non sterile (LaboPlast Bio) or gamma radiation sterilised (SteriPlast Bio)
- Available in two sizes: 2,5 ml (similar to teaspoon) and 10 ml (similar to tablespoon)
- Produced in a cleanroom environment
- Individually packed
They are ideal for the sampling of powder, granules and pastes.
Ordering information: Also available with long handle (11.8") and curved bowl in different sizes - 5 ml (0,17 oz.), 10 ml (0.68 oz.) or 10 ml (0.34 oz.).
Customers Who Bought This Also Bought