LaboPlast® and SteriPlast® Disposable Sampling Spoons, Bürkle
Fournisseur: BURKLE INC
Certificats
A propos de cet article
The LaboPlast® and SteriPlast® single-use spoons made of polystyrene.
- Production, assembling, and packaging acc. to ISO cleanroom class 7
- According to EU food and FDA guidelines
- Individually packed
The spoons are ideal for sampling powders, granules, and pastes.
Ordering information: The sampling spoon is available in 2.5 ml (0.08 oz.) (similar to teaspoon) and 10 ml (0.34 oz.) (similar to tablespoon). Also available with long handle (11.8") and curved bowl in different sizes - 5 ml (0,17 oz.), 10 ml (0.34 oz.) or 20 ml (0.68 oz.).
Customers Who Bought This Also Bought