About this item
Huber Unichillers offer better efficiency, stable pressure and delivery performance with a constant cooling water temperature compared to traditional tap water cooling
Unichillers have the following advantages and functions: environmentally-friendly, economic cooling solution, high efficiency, precise temperature control, powerful circulation pump, robust stainless construction, reliable continuously operation with warning functions, easy to fill and empty, illuminated filling level indicator, LED temperature display, easy operation with only three keys.
Chillers also reduce water consumption and help conserve the environment whilst lowering operating costs. Unichillers are suitable for the removal of heat from chemical processes, cooling technical plants or as a central cooling water supply for laboratories. All Unichillers are designed for continual operation at ambient temperatures up to +40°C. Installation in an outside areas is possible with optional weather protection and winter operation packages. Pumps are available with an extended pressure and flow rate for applications with a high pressure drops.