Multi-Use Test Tubes, Globe Scientific
Fournisseur: Globe Scientific
Certificats
A propos de cet article
Globe Scientific offers an extensive line of multi-use test tubes.
- Polystyrene tubes
- Withstand 1500 rpm during centrifugation
Tubes are molded from premium grade polystyrene for clarity. Tubes are designed to fit most laboratory applications.
Customers Who Bought This Also Bought