ContactUsLinkComponent
Floating Magnet Set, United Scientific Supplies
Floating Magnet Set, United Scientific Supplies
# de catalogue 470144-300
FLOATING MAGNET 6 RING MAGNETS 4 POLE
Floating Magnet Set, United Scientific Supplies
# de catalogue 470144-300
Spécifications
  • Description:
    Set, Floating Magnet
  • Cat. no.:
    470144-300
  • Supplier No.:
    CMSET6
Spécifications
A propos de cet article
Vividly demonstrate the power of magnetic attraction.
  • Sturdy support stand
  • Colorful magnets
  • Easy to demonstrate magnetic repulsion
A special stand has been designed to replace the pencil, to enable hands-free demonstration. Magnets are supplied in plastic cases to reduce breakage. The set contains six brightly colored magnetic rings and the plastic stand, packaged in a cardboard storage box. This set is ideal for demonstrating the laws of magnetic attraction and repulsion. Suitable for ages 11 and above.
Customers Who Bought This Also Bought