To order chemicals, medical devices, or other restricted products please provide ID that includes your business name & shipping address via email [email protected] or fax 484.881.5997 referencing your VWR account number. Acceptable forms of ID are:
- • State issued document with your organization's Federal Tax ID Number
- • State issued document with your organization's Resale Tax ID Number
- • City or County issued Business License
- • State Department of Health Services License
- • Any other ID issued by the State that includes the business name & address
* ATTN: California Customers may require additional documentation as part of the CA Health & Safety Code. Products that fall under this regulation will be placed on a mandatory 21-day hold after documentation is received. VWR will not lift restrictions for residential shipping addresses.
Spécifications
- Antibody type:Primary
- Antigen name:ATG7 autophagy related 7 homolog (S. cerevisiae)
- Clonality:Polyclonal
- Gene ID:O95352
- Host:Rabbit
- Isotype:IgG
- Reactivity:Human,Mouse
- Antigen symbol:ATG7
- Conjugation:Unconjugated
- ELISA:Yes
- ImmunoFluorescence:Yes
- Size:0.02 mg
- Western blot:Yes
- Epitope:near the N-Terminus
- Cross adsorption:No
- Form:Liquid
- Antigen synonyms:ATG7
- Storage buffer:APG7 Antibody is supplied in PBS containing 0.02% sodium azide.
- Molecular weight:Predicted: 77 kDa Observed: 77 kDa
- Storage temperature:APG7 antibody can be stored at 4 °C for three months and –20 °C, stable for up to one year. As with all antibodies care should be taken to avoid repeated freeze thaw cycles. Antibodies should not be exposed to prolonged high temperatures.
- Concentration:1 mg/mL
- Shipping temperature:4 °C
- Immunogen:APG7 antibody was raised against a 17 amino acid synthetic peptide from near the amino terminus of human APG7. The immunogen is located within the first 50 amino acids of APG7.
- Tested applications:Immunocytochemistry
- Purification:APG7 Antibody is affinity chromatography purified via peptide column.
- Cat. no.:10749-734
- Supplier No.:3617_S
Spécifications
A propos de cet article
APG7 Antibody: Autophagy, the process of bulk degradation of cellular proteins through an autophagosomic-lysosomal pathway is important for normal growth control and may be defective in tumor cells. It is involved in the preservation of cellular nutrients under starvation conditions as well as the normal turnover of cytosolic components. This process is negatively regulated by TOR (Target of rapamycin) through phosphorylation of autophagy protein APG1. Another member of the autophagy family of proteins is APG7 which was identified in yeast as a ubiquitin-E1-like enzyme; this function is conserved in the mammalian homolog. In mammalian cells, APG7 is essential for autophagy conjugation systems, autophagosome formation, starvation-induced bulk degradation of proteins and organelles. It has been suggested that caspase-8 may alter APG7 levels and thus the APG7 program of autophagic cell death.
Type: Primary
Antigen: ATG7
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse