PYREX® Drying Tube, [ST] 7/10M
# de catalogue 89091-576
Fournisseur: Corning
Spécifications
- Description:PYREX® Drying Tube, [ST] 7/10M
- Cat. no.:89091-576
- Supplier No.:9090-7
Spécifications
A propos de cet article
Drying tube is a replacement part for the 6949M series of PYREX® microchemistry kits.
Customers Who Bought This Also Bought