Cleanroom Labels, Gloss
Fournisseur: Texwipe
New Product
Certificats
A propos de cet article
The high-energy glossy print skin enhances preprinted labels and thermal transfer inks.
- Removable adhesive
- Made of PP
- Designed for thermal transfer printing
- Use in a cleanroom environment
Available with two different adhesive levels to provide excellent adhesion to a variety of surfaces without leaving residue. Ideal for applications where ink adhesion and reflectivity are desired.
Customers Who Bought This Also Bought