Autoclavable Labels, Texwipe
# de catalogue 82013-940
Fournisseur: Texwipe
Spécifications
- Description:Autoclavable label, blank
- Cat. no.:82013-940
- Packaging:500 labels/roll
- Dimensions:51×51 mm (2×2")
- Supplier No.:TX522A
Spécifications
A propos de cet article
This blank autoclave adhesive labels are designed with pre-printed sterilization indicator.
- Indicator strip turns from blue to pink
Customers Who Bought This Also Bought