ENG Box Electricity
# de catalogue 470165-022
Fournisseur: KIDDER MFG. CO. LTD
Spécifications
- Description:ENG Box Electricity
- Cat. no.:470165-022
- Supplier No.:8354ENGBXE
Spécifications
A propos de cet article
This kit on its own allows a student to experiment with series and parallel circuits, switches and solar power and potentially build their own board game that utilizes the led’s and buzzers.
- Experiment with parallel circuits
- Build a board game
- Includes 117 pieces
This assortment of motors, bulbs, sockets and much more comes in a handy divided utility case. Items included are batteries, batter holders, various bulbs, various sockets, buzzers, motors, switches, and hook-up wire. 117 pieces in total.
Customers Who Bought This Also Bought