A propos de cet article
Cleanroom Labels recommended for use in any cleanroom environment.
EZ-Peel Cleanroom Labels feature adhesive-free zones on the left and right sides of each label, making them easy to peel and position while wearing protective gloves. These labels are manufactured from 100% synthetic, nonparticulating, ultra low-outgassing raw materials. These removable labels can be easily removed without leaving behind adhesive residue contamination. Labels can be printed upon with a cleanroom pen or by any standard thermal transfer printer.
CleanMark® labels can be custom constructed to fit any size, and can be used with either direct thermal or thermal transfer printers. These labels can withstand extreme hot or cold temperatures, harsh chemical solvents, and physical abrasion. They can indicate sterilization processes such as autoclaving, may be pre-sterilized, can firmly adhere to or easily peel from challenging surfaces, and will not contaminate clean or sterile environments with particulates, outgassing, or microbes. Please call your VWR Sales Representative for additional information.