VWR® Freezer Box for Centrifuge Tubes
Catalog # 10002-734
Supplier: VWR International
Specifications
- Color:Clear, Blue Base
- Description:VWR® Freezer Box for Centrifuge Tubes
- Cat. no.:10002-734
Specifications
About this item
VWR® Freezer Box for Centrifuge Tubes features polycarbonate construction and is supplied with lid.
- Suitable over a range of temperatures
- Polycarbonate construction
The box is suitable for use in temperatures as low as -190°C (-310°F). Each box will accommodate 25 x 5 mL centrifuge tubes.
Customers Who Bought This Also Bought