Eye Dropper Bulb
# de catalogue 470205-484
Fournisseur: Walter Stern
Spécifications
- Description:Eye Dropper Bulb
- Cat. no.:470205-484
- Supplier No.:200-2A
Spécifications
A propos de cet article
Latex eye dropper bulb. 2 cc (1.2 oz.) capacity, 0.25" (6mm) bore size, 0.625" (18 mm) diameter, 1.75" (45 mm) length.
Customers Who Bought This Also Bought