Order Entry
ContactUsLinkComponent
Human Recombinant Serpin E2 (from HEK293 Cells)
Human Recombinant Serpin E2 (from HEK293 Cells)
# de catalogue 10798-350
Fournisseur:  Prosci
Numéro CAS:  
Serpin E2 Recombinant Protein
Human Recombinant Serpin E2 (from HEK293 Cells)
# de catalogue 10798-350
Fournisseur:  Prosci
Numéro CAS:  
Restricted Products: To process your orders without delay, please provide the required business documentation to purchase this product.

To order chemicals, medical devices, or other restricted products please provide ID that includes your business name & shipping address via email [email protected] or fax 484.881.5997 referencing your VWR account number. Acceptable forms of ID are:

  • • State issued document with your organization's Federal Tax ID Number
  • • State issued document with your organization's Resale Tax ID Number
  • • City or County issued Business License
  • • State Department of Health Services License
  • • Any other ID issued by the State that includes the business name & address

* ATTN: California Customers may require additional documentation as part of the CA Health & Safety Code. Products that fall under this regulation will be placed on a mandatory 21-day hold after documentation is received. VWR will not lift restrictions for residential shipping addresses.

Spécifications

  • Protein/peptide type:
    Recombinant
  • Source:
    HEK293 cells
  • Species:
    Human
  • Size:
    0.025 mg
  • Tag sequence:
    His Tag
  • Biological activity:
    Measured by its ability to inhibit trypsin cleavage of a fluorogenic peptide substrate, Mca - RPKPVE - Nval - WRK(Dnp) - NH2.
  • Protein synonyms:
    Glia-derived nexin|PI7|SERPINE2|GDN|SerpinE2|Peptidase inhibitor 7|PN-1|Serpin E2|PI-7|Protease nexin 1
  • Protein/peptide name:
    Serpin E2
  • Purity:
    >92% as determined by SDS-PAGE.
  • Molecular weight:
    42.7 kDa
  • Formulation:
    Lyophilized, 20 mM NaAc, 100 mM NaCl, PH6.5
  • Tested applications:
    Western Blot
  • Cat. no.:
    10798-350
  • Supplier No.:
    96-683

Spécifications

A propos de cet article

SERPINE2 is also known as Glia-derived nexin (GDN), Peptidase inhibitor 7 (PI7), Protease nexin 1(PN1). SERPINE2 is a secreted glycoprotein which belongs to the serpin family. SerpinE1 is the primary physiological inhibitor of the two plasminogen activators urokinase (uPA) and tissue plasminogen activator (tPA). PAI-1 / GDN is also implicated in adipose tissue development. It suggests that PAI-1 inhibitors serve in the control of atherothrombosis. Defects in Serpin E1 / PN1 are the cause of plasminogen activator inhibitor-1 deficiency (PAI-1 deficiency) which is characterized by abnormal bleeding due to SerpinE1 defect in the plasma.

Customers Who Bought This Also Bought