Low Ash Quantitative Paper Grade 392, Ahlstrom
Supplier: Ahlstrom
Certificates
About this item
Ahlstrom Grade 392 is a medium rate filtering low ash (Ash 0.01%) filter paper suitable for routine quantitative gravimetric techniques.
It is used for filtration of a wide variety of particles.
Specifications
- Catalog No:
- 10754-472
- 10754-470
- 10754-454
- 10754-476
- 10754-474
- 10754-458
- 10754-456
- 10754-478
- 10754-462
- 10754-460
- 10754-466
- 10754-464
- 10754-468
- Flow rate:
- Medium
- Medium
- Medium
- Medium
- Medium
- Medium
- Medium
- Medium
- Medium
- Medium
- Medium
- Medium
- Medium
- Load capacity:
- Medium
- Medium
- Medium
- Medium
- Medium
- Medium
- Medium
- Medium
- Medium
- Medium
- Medium
- Medium
- Medium
- Retention:
- 5-8 microns
- 5-8 microns
- 5-8 microns
- 5-8 microns
- 5-8 microns
- 5-8 microns
- 5-8 microns
- 5-8 microns
- 5-8 microns
- 5-8 microns
- 5-8 microns
- 5-8 microns
- 5-8 microns