Low Ash Quantitative Paper Grade 391, Ahlstrom
Fournisseur: Ahlstrom
Certificats
A propos de cet article
Ahlstrom Grade 391 is a fine retention low ash (Ash 0.01%) filter paper suitable for routine quantitative gravimetric techniques.
It is used for filtration of fine particles and has a slow flow rate as a result.
Spécifications
- Catalog No:
- 10754-450
- 10754-452
- 10754-440
- 10754-444
- 10754-510
- 10754-508
- 10754-442
- 10754-448
- 10754-514
- 10754-512
- 10754-446
- 10754-518
- 10754-516
- Flow rate:
- Slow
- Slow
- Slow
- Slow
- Slow
- Slow
- Slow
- Slow
- Slow
- Slow
- Slow
- Slow
- Slow
- Load capacity:
- Medium
- Medium
- Medium
- Medium
- Medium
- Medium
- Medium
- Medium
- Medium
- Medium
- Medium
- Medium
- Medium
- Retention:
- 2-3 microns
- 2-3 microns
- 2-3 microns
- 2-3 microns
- 2-3 microns
- 2-3 microns
- 2-3 microns
- 2-3 microns
- 2-3 microns
- 2-3 microns
- 2-3 microns
- 2-3 microns
- 2-3 microns