To order chemicals, medical devices, or other restricted products please provide ID that includes your business name & shipping address via email [email protected] or fax 484.881.5997 referencing your VWR account number. Acceptable forms of ID are:
- • State issued document with your organization's Federal Tax ID Number
- • State issued document with your organization's Resale Tax ID Number
- • City or County issued Business License
- • State Department of Health Services License
- • Any other ID issued by the State that includes the business name & address
* ATTN: California Customers may require additional documentation as part of the CA Health & Safety Code. Products that fall under this regulation will be placed on a mandatory 21-day hold after documentation is received. VWR will not lift restrictions for residential shipping addresses.
Spécifications
- Antibody type:Primary
- Antigen name:Tubulin Alpha 3E
- Clonality:Monoclonal
- Clone:2B4
- Gel shift:Yes
- Gene ID:112714
- Host:Mouse
- Isotype:IgG1
- Reactivity:Human
- Antigen symbol:TUBA3E
- Conjugation:Unconjugated
- Flow cytometry:Yes
- ImmunoFluorescence:Yes
- Size:100 μl
- Western blot:Yes
- Antigen synonyms:p31|HEL-S-91n|P62DOK|Rpn12|Nin1p|HYPF|S14|HIP6
- Storage buffer:PBS (PH 7.3) containing 1% BSA, 50% glycerol and 0.02% sodium azide.
- Storage temperature:For short-term storage, store at 4 °C or store at –20 °C. For long-term storage, store at -70°C (aliquotted). Avoid repeated freeze-thaw cycles.
- Immunogen:Full length human recombinant protein of human TUBA3E (NP_997195) produced in HEK293T cell.
- Purification:Purified from mouse ascites fluids by Protein A/G affinity chromatography
- Cat. no.:89271-740
- Supplier No.:GTX83466
Spécifications
A propos de cet article
Mouse monoclonal antibody [2B4] to TUBA3E
Recommended Starting Dilutions: WB 1:2000 ICC/IF 1:100 Flow Cytometry 1:100Optimal working dilutions should be determined experimentally by the end user.
Type: Primary
Antigen: TUBA3E
Clonality: Monoclonal
Clone: 2B4
Conjugation:
Epitope:
Host: Mouse
Isotype:
Reactivity: Human