To order chemicals, medical devices, or other restricted products please provide ID that includes your business name & shipping address via email [email protected] or fax 484.881.5997 referencing your VWR account number. Acceptable forms of ID are:
- • State issued document with your organization's Federal Tax ID Number
- • State issued document with your organization's Resale Tax ID Number
- • City or County issued Business License
- • State Department of Health Services License
- • Any other ID issued by the State that includes the business name & address
* ATTN: California Customers may require additional documentation as part of the CA Health & Safety Code. Products that fall under this regulation will be placed on a mandatory 21-day hold after documentation is received. VWR will not lift restrictions for residential shipping addresses.
Spécifications
- Antibody type:Primary
- Antigen name:Tubulin Beta 3
- Clonality:Monoclonal
- Clone:2E9
- Gel shift:Yes
- Gene ID:10381
- Host:Mouse
- Isotype:IgG1
- Reactivity:Human
- Antigen symbol:TUBB3
- Conjugation:Unconjugated
- ELISA:Yes
- Flow cytometry:Yes
- ImmunoFluorescence:Yes
- Size:100 μl
- Western blot:Yes
- Antigen synonyms:Tubulin beta-3 chain|CDCBM|beta-4|CFEOM3A|FEOM3|CDCBM1|TUBB4|CFEOM3
- Storage buffer:Ascitic fluid containing 0.03% sodium azide.
- Storage temperature:Store at 4℃, for long term storage, store at -20℃.
- Immunogen:The immunogen corresponding to human TUBB3 expressed in E. Coli.
- Cat. no.:89287-666
- Supplier No.:GTX83356
Spécifications
A propos de cet article
Mouse monoclonal antibody [2E9] to beta III Tubulin
Recommended Starting Dilutions:ELISA: 1/10000.WB: 1/500 - 1/2000.ICC: 1/200 - 1/1000.FCM: 1/200 - 1/400.Not yet tested in other applications.Determining optimal working dilutions by titration test.
Type: Primary
Antigen: TUBB3
Clonality: Monoclonal
Clone: 2E9
Conjugation:
Epitope:
Host: Mouse
Isotype:
Reactivity: Human