Anti-Tubb3 Mouse Monoclonal Antibody [clone: AA10]
# de catalogue 89283-950
Fournisseur: Genetex
Restricted Products: To process your orders without delay, please provide the required business documentation to purchase this product.
To order chemicals, medical devices, or other restricted products please provide ID that includes your business name & shipping address via email [email protected] or fax 484.881.5997 referencing your VWR account number. Acceptable forms of ID are:
- • State issued document with your organization's Federal Tax ID Number
- • State issued document with your organization's Resale Tax ID Number
- • City or County issued Business License
- • State Department of Health Services License
- • Any other ID issued by the State that includes the business name & address
* ATTN: California Customers may require additional documentation as part of the CA Health & Safety Code. Products that fall under this regulation will be placed on a mandatory 21-day hold after documentation is received. VWR will not lift restrictions for residential shipping addresses.
Spécifications
- Antibody type:Primary
- Antigen name:beta III Tubulin
- Clonality:Monoclonal
- Clone:AA10
- Gene ID:246118
- Host:Mouse
- Isotype:IgG
- Reactivity:Rat
- Antigen symbol:TUJ1 (tubulin beta 3)
- Conjugation:Unconjugated
- ImmunoChemistry:Yes
- Size:100 μl
- Western blot:Yes
- Storage buffer:100 µl in 10 mM HEPES (pH 7.5), 150 mM NaCl, 100 µg per ml BSA and 50% glycerol.
- Storage temperature:For long-term storage, aliquot and store at -20 C. Avoid repeated freeze/thaw cycles.
- Concentration:1mg/ml
- Immunogen:Synthetic peptide corresponding to amino acid residues specific to beta III tubulin conjugated to KLH
- Purification:Protein G purified
- Cat. no.:89283-950
- Supplier No.:GTX82572
Spécifications
A propos de cet article
Mouse monoclonal antibody [AA10] to beta III Tubulin
Type: Primary
Antigen: Tubb3
Clonality: Monoclonal
Clone: AA10
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype: IgG
Reactivity: Mouse, Rat
Customers Who Bought This Also Bought