To order chemicals, medical devices, or other restricted products please provide ID that includes your business name & shipping address via email [email protected] or fax 484.881.5997 referencing your VWR account number. Acceptable forms of ID are:
- • State issued document with your organization's Federal Tax ID Number
- • State issued document with your organization's Resale Tax ID Number
- • City or County issued Business License
- • State Department of Health Services License
- • Any other ID issued by the State that includes the business name & address
* ATTN: California Customers may require additional documentation as part of the CA Health & Safety Code. Products that fall under this regulation will be placed on a mandatory 21-day hold after documentation is received. VWR will not lift restrictions for residential shipping addresses.
Spécifications
- Antibody type:Primary
- Antigen name:Cerberus 1, DAN family BMP antagonist
- Clonality:Polyclonal
- Gel shift:Yes
- Gene ID:9350
- Host:Rabbit
- Isotype:IgG
- Reactivity:Human
- Antigen symbol:CER1
- Conjugation:Unconjugated
- ELISA:Yes
- Flow cytometry:Yes
- ImmunoChemistry:Yes
- Size:400 μl
- Western blot:Yes
- Antigen synonyms:CER1|MGC119895|DAND4|MGC119894|MGC96951
- Storage buffer:PBS with 0.09% (W/V) sodium azide
- Storage temperature:Store at at 2-8°C for up to 6 months. For long term storage, aliquot and store at -20 °C. Avoid repeated freeze/thaw cycles.
- Concentration:0.25mg/ml
- Immunogen:KLH conjugated synthetic peptide between 63~93 amino acids from the N-terminal region of human CER1.
- Purification:Prepared by Saturated Ammonium Sulfate (SAS) precipitation followed by dialysis against PBS.
- Cat. no.:89302-350
- Supplier No.:GTX81632
Spécifications
A propos de cet article
Rabbit polyclonal to CER1 (N-term)
Recommended Starting Dilutions:ELISA: Use at 1:1,000WB: Use at 1:50-100IHC: Use at 1:50-100FACS: Use at 1:10-50Optimal dilutions should be determined experimentally by the researcher.
Type: Primary
Antigen: CER1
Clonality: Polyclonal
Clone: 0
Conjugation:
Epitope:
Host: Rabbit
Isotype:
Reactivity: