Anti-CD40L Mouse Monoclonal Antibody [clone: MK13A4]
# de catalogue 89263-470
Fournisseur: Genetex
Restricted Products: To process your orders without delay, please provide the required business documentation to purchase this product.
To order chemicals, medical devices, or other restricted products please provide ID that includes your business name & shipping address via email [email protected] or fax 484.881.5997 referencing your VWR account number. Acceptable forms of ID are:
- • State issued document with your organization's Federal Tax ID Number
- • State issued document with your organization's Resale Tax ID Number
- • City or County issued Business License
- • State Department of Health Services License
- • Any other ID issued by the State that includes the business name & address
* ATTN: California Customers may require additional documentation as part of the CA Health & Safety Code. Products that fall under this regulation will be placed on a mandatory 21-day hold after documentation is received. VWR will not lift restrictions for residential shipping addresses.
Spécifications
- Antibody type:Primary
- Antigen name:CD40 Ligand
- Clonality:Monoclonal
- Clone:MK13A4
- Gene ID:959
- Host:Mouse
- Isotype:IgG1
- Reactivity:Human
- Antigen symbol:CD40L
- Conjugation:Unconjugated
- Flow cytometry:Yes
- Size:50 μg
- Antigen synonyms:hCD40L|TRAP|CD40L|IMD3|gp39|CD154|IGM|T-BAM|TNFSF5|HIGM1|CD40 ligand
- Storage buffer:Phosphate buffered saline
- Storage temperature:Store at –20 °C. This product should be stored undiluted. Storage in frost free freezers is not recommended. Should this product contain a precipitate we recommend microcentrifugation before use.
- Cat. no.:89263-470
- Supplier No.:GTX75664
Spécifications
A propos de cet article
Mouse monoclonal antibody [MK13A4] to CD40L
Type: Primary
Antigen: CD40L
Clonality: Monoclonal
Clone: MK13A4
Conjugation:
Epitope:
Host: Mouse
Isotype:
Reactivity:
Customers Who Bought This Also Bought