To order chemicals, medical devices, or other restricted products please provide ID that includes your business name & shipping address via email [email protected] or fax 484.881.5997 referencing your VWR account number. Acceptable forms of ID are:
- • State issued document with your organization's Federal Tax ID Number
- • State issued document with your organization's Resale Tax ID Number
- • City or County issued Business License
- • State Department of Health Services License
- • Any other ID issued by the State that includes the business name & address
* ATTN: California Customers may require additional documentation as part of the CA Health & Safety Code. Products that fall under this regulation will be placed on a mandatory 21-day hold after documentation is received. VWR will not lift restrictions for residential shipping addresses.
Spécifications
- Antibody type:Primary
- Antigen name:CD48 molecule
- Clonality:Monoclonal
- Clone:MRC OX-45
- Gene ID:245962
- Host:Mouse
- Isotype:IgG1
- Reactivity:Rat
- Antigen symbol:CD48
- Conjugation:Unconjugated
- Flow cytometry:Yes
- Size:2 ml
- Antigen synonyms:MEM-102|BLAST|hCD48|mCD48|BLAST1|BCM1|SLAMF2
- Storage buffer:Phosphate-buffered saline containing 0.02% sodium azide
- Storage temperature:Keep as concentrated solution. Store at 4 °C short term. For extended storage aliquot and store at –20 °C or below. Avoid freeze-thaw cycles.
- Concentration:1.0 mg/ml
- Immunogen:Rat T-blasts prepared by stimulating purified T-helper cells with allogeneic irradiated rat spleen cells.
- Purification:Protein G purified
- Cat. no.:89285-148
- Supplier No.:GTX74635
Spécifications
A propos de cet article
Mouse Antibody to Rat CD48
For FACS: Use at an assay dependent dilution. Not tested in other applications. Optimal dilutions/concentrations should be determined by the researcher.
Type: Primary
Antigen: CD48
Clonality: Monoclonal
Clone: MRC OX-45
Conjugation:
Epitope:
Host: Mouse
Isotype:
Reactivity: