To order chemicals, medical devices, or other restricted products please provide ID that includes your business name & shipping address via email [email protected] or fax 484.881.5997 referencing your VWR account number. Acceptable forms of ID are:
- • State issued document with your organization's Federal Tax ID Number
- • State issued document with your organization's Resale Tax ID Number
- • City or County issued Business License
- • State Department of Health Services License
- • Any other ID issued by the State that includes the business name & address
* ATTN: California Customers may require additional documentation as part of the CA Health & Safety Code. Products that fall under this regulation will be placed on a mandatory 21-day hold after documentation is received. VWR will not lift restrictions for residential shipping addresses.
Spécifications
- Antibody type:Primary
- Antigen name:Interleukin 7
- Clonality:Polyclonal
- Gene ID:16196
- Host:Rabbit
- Isotype:IgG
- Neutralisation:Yes
- Reactivity:Mouse
- Antigen symbol:IL7
- Conjugation:Unconjugated
- ELISA:Yes
- Size:50 μg
- Western blot:Yes
- Antigen synonyms:IL-7|IL7|Interluekin-7|Interluekin 7|IL 7|interluekin
- Storage buffer:Phosphate buffered saline. For long term storage, the addition of 0. 09% sodium azide is recommended. For functional studies, do not use azide.
- Storage temperature:Prior to reconstitution store at +4 °C. Following reconstitution store at –20 °C. Store undiluted. Storage in frost-free freezers is not recommended. Should this product contain a precipitate we recommend microcentrifugation before use.
- Immunogen:Recombinant mouse IL7
- Purification:Purified IgG was prepared from whole serum by affinity chromatography.
- Cat. no.:89263-210
- Supplier No.:GTX74065
Spécifications
A propos de cet article
Rabbit polyclonal antibody to IL7
Recommended Starting Dilutions: ELISA: Max Dilution 0.5 ug/ml Functional Assays: 0.08 ug/ml - 0.1 ug/ml Western Blotting: 0.1 ug/ml - 0.2 ug/ml Not tested in other applications. Optimal dilution / concentration should be determined by the end user.
Type: Primary
Antigen: IL7
Clonality: Polyclonal
Clone:
Conjugation:
Epitope:
Host: Rabbit
Isotype:
Reactivity: