To order chemicals, medical devices, or other restricted products please provide ID that includes your business name & shipping address via email [email protected] or fax 484.881.5997 referencing your VWR account number. Acceptable forms of ID are:
- • State issued document with your organization's Federal Tax ID Number
- • State issued document with your organization's Resale Tax ID Number
- • City or County issued Business License
- • State Department of Health Services License
- • Any other ID issued by the State that includes the business name & address
* ATTN: California Customers may require additional documentation as part of the CA Health & Safety Code. Products that fall under this regulation will be placed on a mandatory 21-day hold after documentation is received. VWR will not lift restrictions for residential shipping addresses.
Spécifications
- Antibody type:Primary
- Antigen name:Interleukin 3
- Clonality:Polyclonal
- Gene ID:16187
- Host:Rabbit
- Isotype:IgG
- Neutralisation:Yes
- Reactivity:Mouse
- Antigen symbol:IL3
- Conjugation:Unconjugated
- ELISA:Yes
- Size:50 μg
- Western blot:Yes
- Antigen synonyms:Multipotential colony-stimulating factor|P-cell-stimulating factor|MCGF|Hematopoietic growth factor|Mast-cell growth factor|Interleukin-3|IL3
- Storage buffer:Phosphate buffered saline
- Storage temperature:Prior to reconstitution store at +4 °C. Following reconstitution store at –20 °C. Store undiluted. Storage in frost-free freezers is not recommended. Should this product contain a precipitate we recommend microcentrifugation before use.
- Immunogen:Highly purified recombinant murine IL3
- Purification:Purified IgG was prepared by antigen affinity chromatography.
- Cat. no.:89263-168
- Supplier No.:GTX74036
Spécifications
A propos de cet article
Rabbit polyclonal antibody to IL3
Recommended Starting Dilutions: ELISA: Max Dilution 0.5 ug/ml Functional Assays: 0.15 ug/ml - 0.20 ug/ml Western Blotting: 0.1 ug/ml - 0.2 ug/ml Not tested in other applications. Optimal dilution / concentration should be determined by
Type: Primary
Antigen: IL3
Clonality: Polyclonal
Clone:
Conjugation:
Epitope:
Host: Rabbit
Isotype:
Reactivity: