Anti-TUBB3 Rabbit Polyclonal Antibody
# de catalogue 89306-718
Fournisseur: Genetex
Restricted Products: To process your orders without delay, please provide the required business documentation to purchase this product.
To order chemicals, medical devices, or other restricted products please provide ID that includes your business name & shipping address via email [email protected] or fax 484.881.5997 referencing your VWR account number. Acceptable forms of ID are:
- • State issued document with your organization's Federal Tax ID Number
- • State issued document with your organization's Resale Tax ID Number
- • City or County issued Business License
- • State Department of Health Services License
- • Any other ID issued by the State that includes the business name & address
* ATTN: California Customers may require additional documentation as part of the CA Health & Safety Code. Products that fall under this regulation will be placed on a mandatory 21-day hold after documentation is received. VWR will not lift restrictions for residential shipping addresses.
Spécifications
- Antibody type:Primary
- Antigen name:Tubulin Beta 3
- Clonality:Polyclonal
- Gene ID:10381
- Host:Rabbit
- Reactivity:Human,Mouse
- Antigen symbol:TUBB3
- Conjugation:Unconjugated
- Size:100 μl
- Western blot:Yes
- Antigen synonyms:Tubulin beta-3 chain|CDCBM|beta-4|CFEOM3A|FEOM3|CDCBM1|TUBB4|CFEOM3
- Storage buffer:saline (without Mg2+ and Ca2+), pH 7.4, 150mM NaCl, 0.02% sodium azide and 50% glycerol.
- Storage temperature:Keep as concentrated solution. Aliquot and store at –20 °C or below. Avoid multiple freeze-thaw cycles.
- Concentration:1 mg/ml
- Immunogen:Peptide sequence around aa.442~446(E-E-S-E-A) derived from Human Tubulin beta-III.
- Purification:Antibodies were purified by affinity-chromatography using epitope-specific peptide.
- Cat. no.:89306-718
- Supplier No.:GTX50789
Spécifications
A propos de cet article
Rabbit polyclonal antibody to beta III Tubulin
- Suggested starting dilutions are as follows. WB: 1:500~1:1000. Not yet tested in other applications. Optimal working dilutions should be determined experimentally by the end user.
Type: Primary
Antigen: TUBB3
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype:
Reactivity: Human, Mouse
Customers Who Bought This Also Bought