To order chemicals, medical devices, or other restricted products please provide ID that includes your business name & shipping address via email [email protected] or fax 484.881.5997 referencing your VWR account number. Acceptable forms of ID are:
- • State issued document with your organization's Federal Tax ID Number
- • State issued document with your organization's Resale Tax ID Number
- • City or County issued Business License
- • State Department of Health Services License
- • Any other ID issued by the State that includes the business name & address
* ATTN: California Customers may require additional documentation as part of the CA Health & Safety Code. Products that fall under this regulation will be placed on a mandatory 21-day hold after documentation is received. VWR will not lift restrictions for residential shipping addresses.
Spécifications
- Antibody type:Primary
- Antigen name:Cyclin E1
- Clonality:Polyclonal
- Gene ID:898
- Host:Rabbit
- Reactivity:Human
- Antigen symbol:CCNE1
- Conjugation:Unconjugated
- Size:100 μl
- Western blot:Yes
- Antigen synonyms:G1/S-specific cyclin-E1|cyclin Et|cyclin Es|CCNE|CYCLE,CCNEcyclin Es
- Modification:phospho-Thr395
- Storage buffer:saline (without Mg2+ and Ca2+), pH 7.4, 150mM NaCl, 0.02% sodium azide and 50% glycerol.
- Storage temperature:Keep as concentrated solution. Aliquot and store at –20 °C or below. Avoid multiple freeze-thaw cycles.
- Concentration:1 mg/ml
- Immunogen:Peptide sequence around phosphorylation site of threonine 395 (L-L-T(p)-P-P)derived from Human Cyclin E1
- Purification:Antibodies were purified by affinity-chromatography using epitope-specific phosphopeptide. Non-phospho specific antibodies were removed by chromatogramphy using non-phosphopeptide.
- Cat. no.:89306-586
- Supplier No.:GTX50722
Spécifications
A propos de cet article
Rabbit Polyclonal antibody to Cyclin E1 (Phospho Thr395)
Suggested starting dilutions are as follows. WB: 1:1000. Not yet tested in other applications. Optimal working dilutions should be determined experimentally by the end user.
Type: Primary
Antigen: CCNE1
Clonality: Polyclonal
Clone:
Conjugation:
Epitope:
Host: Rabbit
Isotype:
Reactivity: Human