To order chemicals, medical devices, or other restricted products please provide ID that includes your business name & shipping address via email [email protected] or fax 484.881.5997 referencing your VWR account number. Acceptable forms of ID are:
- • State issued document with your organization's Federal Tax ID Number
- • State issued document with your organization's Resale Tax ID Number
- • City or County issued Business License
- • State Department of Health Services License
- • Any other ID issued by the State that includes the business name & address
* ATTN: California Customers may require additional documentation as part of the CA Health & Safety Code. Products that fall under this regulation will be placed on a mandatory 21-day hold after documentation is received. VWR will not lift restrictions for residential shipping addresses.
Spécifications
- Antibody type:Primary
- Antigen name:CD45RB antigen
- Clonality:Monoclonal
- Clone:MEM-55
- Gene ID:5788
- Host:Mouse
- Isotype:IgG2b
- Reactivity:Human
- Antigen symbol:CD45RB
- Conjugation:Unconjugated
- Flow cytometry:Yes
- ImmunoChemistry:Yes
- ImmunoPrecipitation:Yes
- Size:100 μg
- Western blot:Yes
- Antigen synonyms:CD45R|B220|CD45|GP180|T200|LY5|LCA
- Storage buffer:Phosphate buffered saline, pH 7.4, containing 15mM sodium azide
- Storage temperature:Keep as concentrated solution. Store at 4 °C short term. For extended storage aliquot and store at –20 °C or below. Avoid freeze-thaw cycles.
- Concentration:1 mg/ml
- Immunogen:Tissue / cell preparation (Human). (Thymocytes and T lymphocytes).
- Purification:Protein A purified
- Cat. no.:89317-440
- Supplier No.:GTX28218
Spécifications
A propos de cet article
Mouse monoclonal [MEM - 55] to CD45RB
For FACS: Use at an assay dependant dilution. For IHC-P: Use at an assay dependant dilution. No pre-treatment of tissue sections is essential. For IP: Use at an assay dependant dilution. For WB: Use at an assay dependant dilution. Not tested in other applications. Optimal dilutions/concentrations should be determined by the researcher.
Type: Primary
Antigen: CD45RB
Clonality: Monoclonal
Clone: MEM-55
Conjugation:
Epitope:
Host: Mouse
Isotype:
Reactivity: