To order chemicals, medical devices, or other restricted products please provide ID that includes your business name & shipping address via email [email protected] or fax 484.881.5997 referencing your VWR account number. Acceptable forms of ID are:
- • State issued document with your organization's Federal Tax ID Number
- • State issued document with your organization's Resale Tax ID Number
- • City or County issued Business License
- • State Department of Health Services License
- • Any other ID issued by the State that includes the business name & address
* ATTN: California Customers may require additional documentation as part of the CA Health & Safety Code. Products that fall under this regulation will be placed on a mandatory 21-day hold after documentation is received. VWR will not lift restrictions for residential shipping addresses.
Spécifications
- Antibody type:Primary
- Antigen name:Cyclin E1
- Clonality:Polyclonal
- Gene ID:898
- Host:Rabbit
- Reactivity:Human,Rat,Mouse
- Antigen symbol:CCNE1
- Conjugation:Unconjugated
- ELISA:Yes
- Size:100 μl
- Western blot:Yes
- Antigen synonyms:G1/S-specific cyclin-E1|cyclin Et|cyclin Es|CCNE|CYCLE,CCNEcyclin Es
- Storage buffer:0.01% (w/v) Sodium Azide
- Storage temperature:Store vial at –20 °C prior to opening.? Aliquot contents and freeze at –20 °C or below for extended storage. Avoid cycles of freezing and thawing.
- Concentration:70 mg/ml
- Immunogen:Anti-Cyclin-E Antibody was produced by repeated immunizations with a KLH conjugated peptide corresponding to a region near the carboxy terminus of human Cyclin E.
- Purification:Antiserum
- Cat. no.:89264-724
- Supplier No.:GTX22094
Spécifications
A propos de cet article
ELISA - Suggested dilution 1:2000. Western blot - Suggested dilution 1:200 to 1:500. Optimal titers for other applications should be determined by the researcher.
Type: Primary
Antigen: CCNE1
Clonality: Polyclonal
Clone:
Conjugation:
Epitope:
Host: Rabbit
Isotype:
Reactivity: Human, Mouse, Rat