To order chemicals, medical devices, or other restricted products please provide ID that includes your business name & shipping address via email [email protected] or fax 484.881.5997 referencing your VWR account number. Acceptable forms of ID are:
- • State issued document with your organization's Federal Tax ID Number
- • State issued document with your organization's Resale Tax ID Number
- • City or County issued Business License
- • State Department of Health Services License
- • Any other ID issued by the State that includes the business name & address
* ATTN: California Customers may require additional documentation as part of the CA Health & Safety Code. Products that fall under this regulation will be placed on a mandatory 21-day hold after documentation is received. VWR will not lift restrictions for residential shipping addresses.
Spécifications
- Antibody type:Primary
- Antigen name:Coagulation Factor 13 A1
- Clonality:Monoclonal
- Clone:AC-1A1
- Gene ID:2162
- Host:Mouse
- Isotype:IgG1
- Reactivity:Human
- Antigen symbol:F13A1
- Conjugation:Unconjugated
- ImmunoChemistry:Yes
- Size:500 μl
- Western blot:Yes
- Antigen synonyms:coagulation factor XIII A chain|A1 polypeptide)|protein-glutamine gamma-glutamyltransferase A chain|A subunit|coagulation factor XIII|coagulation factor XIIIa|fibrinoligase|FSF|bA525O21.1 (coagulation factor XIII|factor XIIIa|TGase|A polypeptide|fibrin stabilizing factor|transglutaminase. plasma|transglutaminase A chain
- Storage buffer:Antibody solution containing sodium azide
- Storage temperature:Keep as concentrated solution, aliquot and store at 4°C. Do not freeze.
- Immunogen:Recombinant human protein corresponding to A-subunit of coagulation Factor XIII.
- Purification:Purified IgG
- Cat. no.:89291-606
- Supplier No.:GTX21834
Spécifications
A propos de cet article
For IHC-P Use at a dilution of 1:25-1:50 in an ABC method for 30 minutes at room temperature. For WB: Use at an assay dependent dilution. Keep as concentrated solution. Store at 4oC short term. For extended storage aliquot and store at -20oC or below. Avoid freeze-thaw cycles.
Type: Primary
Antigen: F13A1
Clonality: Monoclonal
Clone: AC-1A1
Conjugation:
Epitope:
Host: Mouse
Isotype:
Reactivity: Human