To order chemicals, medical devices, or other restricted products please provide ID that includes your business name & shipping address via email [email protected] or fax 484.881.5997 referencing your VWR account number. Acceptable forms of ID are:
- • State issued document with your organization's Federal Tax ID Number
- • State issued document with your organization's Resale Tax ID Number
- • City or County issued Business License
- • State Department of Health Services License
- • Any other ID issued by the State that includes the business name & address
* ATTN: California Customers may require additional documentation as part of the CA Health & Safety Code. Products that fall under this regulation will be placed on a mandatory 21-day hold after documentation is received. VWR will not lift restrictions for residential shipping addresses.
Spécifications
- Antibody type:Primary
- Antigen name:Cyclin E1
- Clonality:Monoclonal
- Clone:168
- Gene ID:898
- Host:Mouse
- Isotype:IgG2a
- Reactivity:Monkey,Human
- Antigen symbol:CCNE1
- Conjugation:Unconjugated
- ImmunoPrecipitation:Yes
- Size:100 μg
- Western blot:Yes
- Antigen synonyms:G1/S-specific cyclin-E1|cyclin Et|cyclin Es|CCNE|CYCLE,CCNEcyclin Es
- Storage buffer:Preservative: 0.08% Sodium Azide; Constituents: PBS
- Storage temperature:Keep as concentrated solution. Store at 4 °C short term. For extended storage aliquot and store at –20 °C or below. Avoid freeze-thaw cycles.
- Concentration:1 mg/ml
- Immunogen:Recombinant full length protein (Human).
- Cat. no.:89278-892
- Supplier No.:GTX16122
Spécifications
A propos de cet article
IP: Use at an assay dependent dilution. WB: Use at an assay dependent dilution. Detects a band of approximately 50 kDa. Not tested in other applications. Optimal dilutions/concentrations should be determined by the end user.
Type: Primary
Antigen: CCNE1
Clonality: Monoclonal
Clone: 168
Conjugation:
Epitope:
Host: Mouse
Isotype:
Reactivity: Human, Monkey