To order chemicals, medical devices, or other restricted products please provide ID that includes your business name & shipping address via email [email protected] or fax 484.881.5997 referencing your VWR account number. Acceptable forms of ID are:
- • State issued document with your organization's Federal Tax ID Number
- • State issued document with your organization's Resale Tax ID Number
- • City or County issued Business License
- • State Department of Health Services License
- • Any other ID issued by the State that includes the business name & address
* ATTN: California Customers may require additional documentation as part of the CA Health & Safety Code. Products that fall under this regulation will be placed on a mandatory 21-day hold after documentation is received. VWR will not lift restrictions for residential shipping addresses.
Spécifications
- Antibody type:Primary
- Antigen name:Eotaxin 2 (Chemokine (C-C motif) ligand 24)
- Clonality:Polyclonal
- Gene ID:56221
- Host:Goat
- Isotype:IgG
- Neutralisation:Yes
- Reactivity:Mouse
- Antigen symbol:Ccl24
- Conjugation:Unconjugated
- ELISA:Yes
- Size:50 μg
- Western blot:Yes
- Storage buffer:Phosphate buffered saline
- Storage temperature:May be stored at 4 °C for short-term only. Aliquot to avoid repeated freezing and thawing. Store at –20 °C. Aliquots are stable for at least a year. Centrifuge the original vial after thawing and prior to removing the cap
- Immunogen:Recombinant mouse Eotaxin 2
- Purification:Purified IgG was prepared from whole serum by affinity chromatography.
- Cat. no.:89286-084
- Supplier No.:GTX12462
Spécifications
A propos de cet article
Recommended Starting Dilutions:ELISA: Max Dilution 0.5 ug/mlFunctional Assays: 7.0 - 8.0 ug/mlWestern Blotting: 0.1 - 0.2 ug/mlNot tested in other applications. Optimal dilution / concentration should be determined by the end user.
Type: Primary
Antigen: Ccl24
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Goat
Isotype: IgG
Reactivity: Mouse