To order chemicals, medical devices, or other restricted products please provide ID that includes your business name & shipping address via email [email protected] or fax 484.881.5997 referencing your VWR account number. Acceptable forms of ID are:
- • State issued document with your organization's Federal Tax ID Number
- • State issued document with your organization's Resale Tax ID Number
- • City or County issued Business License
- • State Department of Health Services License
- • Any other ID issued by the State that includes the business name & address
* ATTN: California Customers may require additional documentation as part of the CA Health & Safety Code. Products that fall under this regulation will be placed on a mandatory 21-day hold after documentation is received. VWR will not lift restrictions for residential shipping addresses.
Spécifications
- Antibody type:Primary
- Antigen name:Vacuolar Protein Sorting 45
- Clonality:Monoclonal
- Clone:22A11
- Gene ID:796
- Host:Mouse
- Isotype:IgG1
- Reactivity:Human,Bovine,Dog,Mouse
- Antigen symbol:VPS45
- Conjugation:Unconjugated
- ELISA:Yes
- Size:100 μg
- Western blot:Yes
- Antigen synonyms:H1VPS45|vacuolar protein sorting 45A|H1|VSP45|VPS45A|leucocyte vacuolar protein sorting 45|vacuolar protein sorting 45B (yeast)|vacuolar protein sorting 45A (yeast)|vacuolar protein sorting 45A (yeast homolog)|VPS45B|RP11-458I7.2|VPS54A|VPS45|VSP45A|vacuolar protein sorting 45 homolog (S. cerevisiae)|vacuolar protein sorting protein similar to S. cerevisiae Vsp45p
- Storage buffer:Phosphate-buffered saline, pH 7.4, containing 0.1% sodium azide
- Storage temperature:Keep as concentrated solution, aliquot and store at 4°C.
- Immunogen:Full length protein (Human).
- Purification:Protein A purified
- Cat. no.:89336-544
- Supplier No.:GTX11491
Spécifications
A propos de cet article
ELISA: Use at an assay dependent dilution. WB: Use at an assay dependent dilution. Pair for sandwich ELISA: GTX11498 Procalcitonin antibody [38F11] for capture antibody, and GTX11491 for detection antibody. Optimal dilutions should be determined experimentally by the end user.
Type: Primary
Antigen: VPS45
Clonality: Monoclonal
Clone: 22A11
Conjugation:
Epitope:
Host: Mouse
Isotype:
Reactivity: Human, Mouse, Bovine, Dog